Superfrost® Plus Gold Slides, Electron Microscopy Sciences | Avantor
Superfrost® Plus Gold Slides, Electron Microscopy Sciences
Plus Gold Slide
Catalog # 100501-732
Fournisseur #:  71864-10
Superfrost® Plus Gold Slides, Electron Microscopy Sciences
Catalog # 100501-732
Superfrost® Plus Gold Slides, Electron Microscopy Sciences
Superfrost® Plus Gold Slides, Electron Microscopy Sciences
Plus Gold Slide
Price
$1,296.29
Log in pour la disponibilité
Case of 10 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These slides are specially designed for the application of fresh or formalin fixed frozen tissue. They are inherently tissue binding so there is no need to add sprays or adhesives to the slides.
  • Securely attach frozen breast, brain, skin and other normally difficult to bond tissue sections to the slide
  • Ideal when special stains, immunocytochemical and in situ DNA hybridization techniques are applied on fresh and fixed frozen tissue
  • Assist tissue adherence during immunocytochemical analysis of estrogen and progesterone receptors in fresh frozen breast tissue
  • They are compatible with both toluidine blue and hematoxylin and eosin rapid frozen section stains
Spécifications
  • Désignation:
    Plus Gold Slide