Grid Mats, Electron Microscopy Sciences
Grid Mats, Electron Microscopy Sciences
Square grid mats
Catalog # 100499-708
Supplier #:  71162
Grid Mats, Electron Microscopy Sciences
Catalog # 100499-708
Grid Mats, Electron Microscopy Sciences
Grid Mats, Electron Microscopy Sciences
Square grid mats
Price
$219.34
Log in for availability
Dozen Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
White silicone rubber mats, with numbered compartments.
  • Good for organizing grids
  • They will not slide or jump between compartments
  • Ideal for staining grids
Easy to pick up grids without damaging forcep tips. Mats are available for square and round petri dishes, (100 mm diameter).
Specifications
  • Désignation:
    Square grid mats