Etched Double Neubauer, Electron Microscopy Sciences
Chamber With Two Cover Glasses
  100498-470
 :  63511-11
Etched Double Neubauer, Electron Microscopy Sciences
  100498-470
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Chamber With Two Cover Glasses
$1,266.81
Each
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.
 : The complete set includes one counting chamber, two 0.5 mm cover glasses, pipettes, and carrying case.
  • Désignation:
    Chamber With Two Cover Glasses