Inoculating Loops, Electron Microscopy Sciences
Product Details & Documents
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
Spécifications
- Désignation:Inoculating Loops
- Taille:3 x 70 mm
- Jauge:26 Gauge