SEM Conductive Tapes, Electron Microscopy Sciences
SEM Conductive Tapes, Electron Microscopy Sciences
Double Sided Carbon Tape, 20 mm
  100494-404
Supplier #:  77817-20
SEM Conductive Tapes, Electron Microscopy Sciences
  100494-404
SEM Conductive Tapes, Electron Microscopy Sciences
SEM Conductive Tapes, Electron Microscopy Sciences
Double Sided Carbon Tape, 20 mm
$251.94
Each
We now offer a new line of conductive tapes, that are electrically conductive and have a clean background. They were developed especially for scanning electron microscopy.
  • Désignation:
    Double Sided Carbon Tape, 20 mm