EMS Flat-Tip Tweezers, Electron Microscopy Sciences | Avantor
EMS Flat-Tip Tweezers, Electron Microscopy Sciences
Straight Spade Tip Forcepts, Type 127, 105 mm, Thin and square tips, straight
Catalog # 100493-928
Fournisseur #:  72972-SP
EMS Flat-Tip Tweezers, Electron Microscopy Sciences
Catalog # 100493-928
EMS Flat-Tip Tweezers, Electron Microscopy Sciences
EMS Flat-Tip Tweezers, Electron Microscopy Sciences
Straight Spade Tip Forcepts, Type 127, 105 mm, Thin and square tips, straight
Price
$73.62
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These long and slender steel tweezers work well in multiple precision applications and are especially well suited for working near heat sources.
  • Made from SA steel
  • Serrated grips
Spécifications
  • Désignation:
    Straight Spade Tip Forcepts, Type 127
  • Longueur:
    105 mm
  • Tip Style:
    Thin and square tips, straight