Five-Channel Timer, Electron Microscopy Sciences
Timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours.
- Memory recalls last timer setting for reuse
- Has a clock mode
Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero.
Dimensions: 2⁷/₈×2¹/₂×⁹/₁₆"
Weight: 2 oz.
: Comes complete with a stand, pocket clip, and magnet.
- Désignation:Timer 5-channel certified