Low Temperature Dessicator, Electron Microscopy Sciences
Product Details & Documents
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm (1/4") acrylic with full-length hinges and stainless steel twist locking latches.
- Easy viewing clear units or dark bonze for light sensitive materials
- Deep, slide-in trays with perforated bottom
- Available in three sizes
Spécifications
- Désignation:Low Temperature Desiccator Cabinet Bronze