Embryo Dishes, Electron Microscopy Sciences | Avantor
Embryo Dishes, Electron Microscopy Sciences
Embryo Dish, 45 mm
Catalog # 100492-982
Fournisseur #:  70543-45
Embryo Dishes, Electron Microscopy Sciences
Catalog # 100492-982
Embryo Dishes, Electron Microscopy Sciences
Embryo Dishes, Electron Microscopy Sciences
Embryo Dish, 45 mm
Price
$58.36
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These dishes are molded from clear glass and come with a glass lid for the viewing or staining of free-floating specimens.
Spécifications
  • Ø ext.:
    45 mm
  • Désignation:
    Embryo Dish