Marker Pen, Markal Sharpie, Electron Microscopy Sciences
Product Details & Documents
Smear resistant, water-resistant, fast-drying no
15 fine line. Usable on glass, porcelain, metals, and plastics.
Spécifications
- Désignation:Marker Pen Black