Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Emery refills
Catalog # 100490-142
Supplier #:  62118-12
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Catalog # 100490-142
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Filing/Polishing Stick and Strips, Electron Microscopy Sciences
Emery refills
Price
$63.82
Log in for availability
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A plastic handle, 267 mm (10 1/2") long, holds strips of emery paper or cloth 279 x 25.4 mm (11 x 1"). Strips are easily replaced. Comes complete with 2 strips/set (grit 120 and 240).
Specifications
  • Désignation:
    Emery refills