Cotton Tipped Applicators, Electron Microscopy Sciences
Product Details & Documents
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length, 1/12" Ø wooden shaft and head size is 3/16" (Ø) x 5/8" (L).
Handy Vial Packs: 3" (8 cm) wooden shaft, small-tipped applicators come pre-packed in a plastic-capped vial, a handy way to keep a supply of swabs available anytime on the bench top.
Spécifications
- Sterility:Non sterile
- Désignation:Cotton Tipped Applicator, Single End
- Packaging:100/Pack