Tooled Neck Vials, Electron Microscopy Sciences
Product Details & Documents
These vials are made from Type 1, Class B borosilicate glass. Their tooled necks ensure accurate fit with 'snap-in' closures. They are ideal for storage of specimens and sample blocks.
Delivery information: The 1 and 2 dram sizes come with an open-bottom closure and all of the other sizes come with a closed bottom, two-piece closure.
Packaging: Closures are packed separately.
Specifications
- Avec bouchon:Oui
- Désignation:Tooled Neck Vials, 4DR