Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
Product Details & Documents
Holds up to 5 slides 3x1"in size . These mailers are rigid, strong and durable with a convenient flip-top, snap-on lid.
When it stands in its upright position, it can be used as a staining container, with a minimal amount of solution being used. Packaged 25 mailers/pk, 8 packs/cs.
Spécifications
- Désignation:Model 2