Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Single Microscope Slide Mailer, Solid Cardboard
Catalog # 100489-336
Supplier #:  71539-10
Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Catalog # 100489-336
Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Microscope Slide Mailers, Solid Cardboard, Electron Microscopy Sciences
Single Microscope Slide Mailer, Solid Cardboard
Price
$633.03
Log in pour la disponibilité
Each Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The slide mailers are available in sizes to hold one, two, or four 3 x 1" glass slides.
1-Slide Solid Cardboard Slide Mailer is made from 0.050” (1.29 mm) solid cardboard, measures 33/4 x 15/8” when folded. The mailer holds one 3 x 1” glass slide with or without cover glass and has a thumb groove for easy removal. A special cut corner identifies the top of the mailer.
2-Slide Solid Cardboard Slide Mailer is made from .085” (2.17 mm) cardboard and measures 3 x 41/4” when folded. This mailer is unprinted. Upon request the mailer can be printed with warnings for medical specimens. Each mailer conforms to U.S. Postal regulations regarding minimum size for First Class.
4-Slide Solid Cardboard Slide Mailer has the same construction as 2-slide mailer above but holds 4 of 3 x 1" slide.
Spécifications
  • Désignation:
    Single Microscope Slide Mailer, Solid Cardboard