Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Product Details & Documents
This low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette. The pipettes are graduated for nominal indication of amount of liquid being drawn. Bellows-shaped bulb makes for easy use and provides excellent drop information.
Spécifications
- Capacité:250 ml (8.5 oz.)