Microsurgical Knife, Electron Microscopy Sciences
Microsurgical Knife, Electron Microscopy Sciences
Micro Surgical Knife 30°
Catalog # 101413-488
Supplier #:  72047-30
Microsurgical Knife, Electron Microscopy Sciences
Catalog # 101413-488
Microsurgical Knife, Electron Microscopy Sciences
Microsurgical Knife, Electron Microscopy Sciences
Micro Surgical Knife 30°
Price
$184.43
Log in for availability
Each Pack of 6
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from tool steel wire. Sharpened to microsurgical quality.
The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
Packaging: 6 blister packs to a box.
Specifications
  • Désignation:
    Micro Surgical Knife 30°