Retrieving Tweezers, Electron Microscopy Sciences
Retrieving Tweezers, Electron Microscopy Sciences
Retrieving Tweezers, 10"
Catalog # 101412-602
Supplier #:  62112-10
Retrieving Tweezers, Electron Microscopy Sciences
Catalog # 101412-602
Retrieving Tweezers, Electron Microscopy Sciences
Retrieving Tweezers, Electron Microscopy Sciences
Retrieving Tweezers, 10"
Price
$41.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A unique plastic coating on the tips of the tweezers insulates them and provides resistance to heat and chemicals. Ideal when cleaning with ultrasonics.
Spécifications
  • Désignation:
    Retrieving Tweezers
  • Longueur:
    10"