StainTray - Slide Staining System, Electron Microscopy Sciences
StainTray - Slide Staining System, Electron Microscopy Sciences
Black Lid Only for 10 Slide Staining Tray
Catalog # 101413-414
Supplier #:  71398-B
StainTray - Slide Staining System, Electron Microscopy Sciences
Catalog # 101413-414
StainTray - Slide Staining System, Electron Microscopy Sciences
StainTray - Slide Staining System, Electron Microscopy Sciences
Black Lid Only for 10 Slide Staining Tray
Price
$177.91
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An ideal device for immunohistochemistry slide staining. The StainTray™ has a black base, made from tough ABS plastic, withstanding a wide range of chemicals (except chlorinated hydrocarbons). It will accept up to 20 slides on four plastic rails covered with a polymer strip to perfectly hold the slides even if the tray is held at an angle.
  • Suitable for routine staining requiring a humid chamber
  • Ideal for hematology, cytology and microscopy laboratories
  • 20-Slide unit with cover: 15 x 93/8 x 13/4" H (38 x 24 x 4.5 cm H)
  • 10-Slide unit with cover: 93/8 x 93/8 x 1" (24 x 24 x 4.5 cm H)
When humidity is needed, the wells between the rails will hold the water securely without splashing. Rails are raised not only to avoid water touching the slides but also make the slides more easily retrievable. The base will also hold excess stain solution that drips from the slides. Four rubber feet add to the base for stability. Units are stackable to save space.
A clear cover is available that allows for visual examination. It is made from PETG with temperature a range of -20 to 60 °C. A black cover is available for fluorescent work. It is made from ABS with a temperature range of -80 to 80 °C.
Specifications
  • Désignation:
    Black Lid Only for 10 Slide Staining Tray