Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences | Avantor
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
500 ml (17 oz.)
Catalog # 100503-278
Fournisseur #:  60960
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
Catalog # 100503-278
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
500 ml (17 oz.)
Price
$144.24
Log in pour la disponibilité
Each Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from heavy gauge polypropylene
  • Resistant to most chemicals
  • Frosted marking area
  • Customary and metric graduations
  • Anti-drip ridge
Suitable for parafilm cover.
Spécifications
  • Capacité:
    500 ml (17 oz.)