Tri-Pour Beaker Caps, Electron Microscopy Sciences
Tri-Pour Beaker Caps, Electron Microscopy Sciences
250 ml (8.5 oz.) beaker
Catalog # 100503-210
Supplier #:  60973
Tri-Pour Beaker Caps, Electron Microscopy Sciences
Catalog # 100503-210
Tri-Pour Beaker Caps, Electron Microscopy Sciences
Tri-Pour Beaker Caps, Electron Microscopy Sciences
250 ml (8.5 oz.) beaker
Price
$176.53
Log in pour la disponibilité
Each Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The caps are wax impregnated, have lift-tabs, and may be written on for sample identification.
The caps are available for all Tri-Pour beakers.
Spécifications
  • Fits:
    250 ml (8.5 oz.) beaker