Needle Probe Set, Electron Microscopy Sciences | Avantor
Needle Probe Set, Electron Microscopy Sciences
Replacement Probe #3, 120 mm
Catalog # 100503-064
Fournisseur #:  62130-03
Needle Probe Set, Electron Microscopy Sciences
Catalog # 100503-064
Needle Probe Set, Electron Microscopy Sciences
Needle Probe Set, Electron Microscopy Sciences
Replacement Probe #3, 120 mm
Price
$15.51
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Set consists of one lightweight precision handle and four interchangeable tools. Can be used for manipulating micro samples, scribing, spring assembly, and probing. The handle is finely balanced, non-rolling gold anodized with split collet and a knurled lock nut. The length is 120 mm (4.7"). Precision forged and ground tools are hardened and nickel plated tool steel with sharp tips.
Spécifications
  • Désignation:
    Replacement Probe #3
  • Taille:
    120 mm