StaticMaster Brushes, Electron Microscopy Sciences
Product Details & Documents
Eliminate static electric charges, and remove dust, lint and other foreign particles in one quick operation. StaticMaster contains Polonium210 element which emits low energy alpha particles. Alpha particles are non-penetrating, stopped completely by a thin sheet of paper or a few inches of air.
- Ionizes air to create a conductive path to remove static
- Static charge is safely and effectively grounded
- Useful life of at least one year
StaticMaster 1C200: This 1" model cleans negatives, mounted or unmounted transparencies, cameras, and smaller enlarging lenses. It is ideal for use with 35 mm or 21/4" x 21/4: film formats. In addition, this model is perfect for removing static and dust from the inside of cameras as well as other equipment. Brush of soft, natural hair removes dust and lint.
StaticMaster 3C500: The 3" model cleans large areas quickly and effectively. Its brush is retractable to just a desired length making it perfect for use with 4 x 5 or smaller film formats. It is also handy for cleaning enlarger condenser lenses, film holders, and other large surfaces. While the active element is 2" wide, in one stroke it actually eliminates static across a 4" area. For cleaning very large areas, such as 8x10" films, two or more 3" StaticMaster may be joined together with easy-to-install StaticMaster Connectors are available separately.
StaticMaster Replacement Cartridges: The StaticMaster has a useful life of at least one year. The element gradually loses its power to neutralize static and should be replaced. For this reason, StaticMasters are stamped with an expiration date. You can easily install a new cartridge without returning the brush to the factory.
StaticMaster Connectors: With this device, you are able to join several 3" models StaticMaster brushes together to make a continuous brush of greater width. For instance, 2 Models 3C500 make 6" brush that can eliminate static over an 8" area. Easy to install and remove.
Specifications
- Désignation:Static Master 3" Replacement Cartridge