Foreline Traps, Electron Microscopy Sciences
Foreline Traps, Electron Microscopy Sciences
Foreline Traps 11/2", 10 CFM, Aluminum
Catalog # 100502-328
Supplier #:  91050-25
Foreline Traps, Electron Microscopy Sciences
Catalog # 100502-328
Foreline Traps, Electron Microscopy Sciences
Foreline Traps, Electron Microscopy Sciences
Foreline Traps 11/2", 10 CFM, Aluminum
Price
$1,045.26
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These rechargeable in-line traps effectively block oil backstreaming from mechanical pumps. Any mechanically pumped system, even one with a turbo pump, should have a foreline traps. The trap is placed in the foreline between the roughing pump and the diffusion or turbo pump. It uses a disposable, oxygen-free copper maze absorbent which requires no liquid nitrogen and will not hold water. Typically the maze is replaced every 1-2 years. The seamless body is available in aluminum. Replacement elements (Copper Maze) also available.
  • A quick change clamp for replacement of maze
  • A viton “O” ring end cap
  • A recessed screen
  • Available in five pipe diameters
Specifications
  • Capacité:
    10 CFM
  • Désignation:
    Foreline Traps 11/2"
  • Matière:
    Aluminum