Light Pipe Scintillators, Electron Microscopy Sciences
Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mm
  100502-320
 :  82000-104
Light Pipe Scintillators, Electron Microscopy Sciences
  100502-320
Light Pipe Scintillators, Electron Microscopy Sciences
Light Pipe Scintillators, Electron Microscopy Sciences
Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mm
$1,650.57
Each
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
  • Scintillator material coated directly on the light pipe which improves the signal
  • Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
 : Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.
  • Désignation:
    Cambridge Light Pipe Scintillator, Quartz, Straight, 104 mm