Glazed Porcelain Plate, Electron Microscopy Sciences | Avantor
Glazed Porcelain Plate, Electron Microscopy Sciences
Glazed Porcelain Plate, 6 Well
Catalog # 100502-186
Fournisseur #:  71575-06
Glazed Porcelain Plate, Electron Microscopy Sciences
Catalog # 100502-186
Glazed Porcelain Plate, Electron Microscopy Sciences
Glazed Porcelain Plate, Electron Microscopy Sciences
Glazed Porcelain Plate, 6 Well
Price
$7.09
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our economical glazed porcelain plate is made from high purity raw material, uniform in quality and resistant to acids and alkalis.
It can withstand sudden temperature changes without cracks, explosion or deformation. Under normal conditions, the glazed plate can sustain a temperature of up to 1050 °C. Available in two models.
Spécifications
  • Désignation:
    Glazed Porcelain Plate, 6 Well