Hi Temperature Indicator Tape, Electron Microscopy Sciences
Product Details & Documents
Specialty cloth-textured Hi Temperature Indicator tape shows the word “STERILIZED” in bold, black letters after standard sterilization processing. The tape sticks to almost any surface or shape, holds fast in 550°F dry oven temperatures. Available in widths of 1/2", 3/4", and 1" on 250” rolls.
Spécifications
- Désignation:Hi-Temp Indicator Tape, 1" Wide