Replacement Desiccator Plates, Electron Microscopy Sciences
140 mm
  100501-418
 :  71242-15
Replacement Desiccator Plates, Electron Microscopy Sciences
  100501-418
Replacement Desiccator Plates, Electron Microscopy Sciences
Replacement Desiccator Plates, Electron Microscopy Sciences
140 mm
$263.51
Each
These 4.8 mm thick plates are able to withstand temperatures up to 150°C (300°F) and have excellent resistance to thermal shock, and to chemicals. They are non-combustible and asbestos free. Round plates are perforated with 4 mm (0.165”) diameter holes on 12.7 mm (1/2”) centers in a straight line pattern. They are available in three sizes, and fit our desiccators.
  • Taille:
    140 mm