Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Gray
Catalog # 100500-738
Supplier #:  70078-GR
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Catalog # 100500-738
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
Gray
Price
$481.17
Log in for availability
Box of 250 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it’s own compartment, numbered from 1 to 6 and measuring 7 x 12 x 5 mm (1/4 x 7/16 x 3/16"). Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45° angle, making the cassette, to some degree, able to be used with certain types of cassette labeling instruments. Each case contains 4 dispenser boxes of 250 cassettes with covers assembled.
Specifications
  • Color:
    Gray