Cryo Markers, Electron Microscopy Sciences
Product Details & Documents
Used for permanently marking cloth, cardboard, and plastic.
It will not fade at ultra-low temperatures. Smudge-proof. Package consist of four, extra fine point pens in assorted colors (red, green, blue, and black).
Specifications
- Désignation:Cryo markers