Flat Emery Sticks, Electron Microscopy Sciences
Flat Emery Sticks, Electron Microscopy Sciences
3 grit emery stick
Catalog # 100499-886
Supplier #:  62119-30
Flat Emery Sticks, Electron Microscopy Sciences
Catalog # 100499-886
Supplier:  
Flat Emery Sticks, Electron Microscopy Sciences
Flat Emery Sticks, Electron Microscopy Sciences
3 grit emery stick
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Emery paper which is glued to a wooden handle. Very useful for filing excess resin of sample block tips. Grit size ranges from number 4/0 (finest) to 3 (coarsest).
Dimensions L x W: 279 x 19.1 mm (11 x 3/4").
Spécifications
  • Désignation:
    3 grit emery stick