Flat Emery Sticks, Electron Microscopy Sciences | Avantor
Flat Emery Sticks, Electron Microscopy Sciences
1 grit emery stick
Catalog # 100499-882
Fournisseur #:  62119-10
Flat Emery Sticks, Electron Microscopy Sciences
Catalog # 100499-882
Flat Emery Sticks, Electron Microscopy Sciences
Flat Emery Sticks, Electron Microscopy Sciences
1 grit emery stick
Price
$60.12
Log in pour la disponibilité
Dozen Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Emery paper which is glued to a wooden handle. Very useful for filing excess resin of sample block tips. Grit size ranges from number 4/0 (finest) to 3 (coarsest).
Dimensions L x W: 279 x 19.1 mm (11 x 3/4").
Spécifications
  • Désignation:
    1 grit emery stick