Dissecting Needles, Electron Microscopy Sciences | Avantor
Dissecting Needles, Electron Microscopy Sciences
Straight Dissecting Needle
Catalog # 100499-712
Fournisseur #:  72948
Dissecting Needles, Electron Microscopy Sciences
Catalog # 100499-712
Dissecting Needles, Electron Microscopy Sciences
Dissecting Needles, Electron Microscopy Sciences
Straight Dissecting Needle
Price
$27.72
Log in pour la disponibilité
Dozen Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The stainless steel with spear shaped head measures 127 mm (5") long. The wooden handle has a needle tip.
Spécifications
  • Désignation:
    Straight Dissecting Needle