Grid Mats, Electron Microscopy Sciences
Product Details & Documents
White silicone rubber mats, with numbered compartments.
- Good for organizing grids
- They will not slide or jump between compartments
- Ideal for staining grids
Easy to pick up grids without damaging forcep tips. Mats are available for square and round petri dishes, (100 mm diameter).
Specifications
- Désignation:Round grid mats