Traceable Calibration Standards, Electron Microscopy Sciences
Traceable Calibration Standards, Electron Microscopy Sciences
PS1 Micrometer Standard with NPL Cert, 2 X 0.05 mm Slot
Catalog # 100499-348
Supplier #:  68047-1NP
Traceable Calibration Standards, Electron Microscopy Sciences
Catalog # 100499-348
Traceable Calibration Standards, Electron Microscopy Sciences
Traceable Calibration Standards, Electron Microscopy Sciences
PS1 Micrometer Standard with NPL Cert, 2 X 0.05 mm Slot
Price
$5,419.97
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These stage micrometers are available with a certificate giving provide traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, which is necessary under ISO provisions.
  • Glass discs are mounted in stainless steel slides with engraved serial numbers
  • Each slide is supplied in a polished wooden storage case to distinguish it as a traceable standard of high value
  • Calibration by NPL, UKAs and Pyser-SGI
The calibration of the scales and grids is carried out by the most appropriate laboratory to suit the customers requirements. The choice of laboratory is normally dependent on the nature of the calibration and the accuracy required.
Calibration by NPL: The National Physical Laboratory carries out measurements at selected points on the scale and grids and issues a certificate of calibration. Order Code NP. The NPL is the English equivalent of NIST.
Calibration by UKAS accredited laboratory: A UKAS accredited laboratory carries out measurements at selected points on the scales and grids and issues a calibration certificate. Order Code: NA
Measurement by Pyser-SGI, Graticules Division: For applications, which do not require the accuracy or traceability provided by calibrations carried out by NPL or a UKAS accredited laboratory, We can provide a Certificate of Comparison. The scale or grid is compared with NPL calibrated in-house standards and a statement is provided on the accuracy of the item with respect to these standards. Order Code: GR
 : Calibration certification is available.
Spécifications
  • Désignation:
    PS1 Micrometer Standard with NPL Cert
  • Taille:
    2 X 0.05 mm Slot