Full Protractor, Electron Microscopy Sciences | Avantor
Full Protractor, Electron Microscopy Sciences
NE45 Eyepiece Scale, 21 mm
Catalog # 100499-114
Fournisseur #:  68002-21
Full Protractor, Electron Microscopy Sciences
Catalog # 100499-114
Full Protractor, Electron Microscopy Sciences
Full Protractor, Electron Microscopy Sciences
NE45 Eyepiece Scale, 21 mm
Price
$357.90
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
NE45 eyepiece scale is surface chrome picture, 10 mm diameter separated in degrees, 16 mm full protractor scale.
Spécifications
  • Désignation:
    NE45 Eyepiece Scale
  • Longueur:
    21 mm