Gold Seal Plain Microscope Slides, Electron Microscopy Sciences
Product Details & Documents
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.
Spécifications
- Capacité:1 g
- Désignation:Plain slides
- L×l:75×25 mm (3×1")
- Épaisseur:1.2 mm