Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Ruled Hemacytometer, Complete Set
Catalog # 100498-468
Supplier #:  63511-10
Etched Double Neubauer, Electron Microscopy Sciences
Catalog # 100498-468
Etched Double Neubauer, Electron Microscopy Sciences
Etched Double Neubauer, Electron Microscopy Sciences
Ruled Hemacytometer, Complete Set
Price
$1,821.36
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Double improved Neubauer rulings are diamond etched directly into a polished recess to reduce damage from scratches. Opaque surfaces above and below the counting area offer an easy focal reference. The chamber comes with a quick orientation mark for the microscope. 0.1 mm cell depth.
Ordering information: The complete set includes one counting chamber, two 0.5 mm cover glasses, pipettes, and carrying case.
Spécifications
  • Désignation:
    Ruled Hemacytometer, Complete Set