Inoculating Loops, Electron Microscopy Sciences | Avantor
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 3 x 70 mm, 24 Gauge
Catalog # 100498-100
Fournisseur #:  62436-03
Inoculating Loops, Electron Microscopy Sciences
Catalog # 100498-100
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, Electron Microscopy Sciences
Inoculating Loops, 3 x 70 mm, 24 Gauge
Price
$560.03
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.
Spécifications
  • Désignation:
    Inoculating Loops
  • Taille:
    3 x 70 mm
  • Jauge:
    24 Gauge