Embedding Blocks, Electron Microscopy Sciences
Embedding Block 1, 19 x 19 x 19 mm
Catalog # 100498-054
 :  62363-01
Embedding Blocks, Electron Microscopy Sciences
Catalog # 100498-054
Embedding Blocks, Electron Microscopy Sciences
Embedding Blocks, Electron Microscopy Sciences
Embedding Block 1, 19 x 19 x 19 mm
Price
$160.14
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.
Spécifications
  • Désignation:
    Embedding Block 1
  • Dimensions:
    19 x 19 x 19 mm