Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
A versatile hang-up flexible shaft machine.
: Mounted accessories are on 2.35 mm (3/32") arbors.
: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
- Désignation:1/8 Hp Flex Shaft Motor
- Tension:220 V