Static Control Adjustable Wrist Band, Electron Microscopy Sciences
Product Details & Documents
This hook and loop wrist strap is designed for those whom need to be static free in the working area.
This hook and loop wrist trap has a quick and easy adjustment. With a 1" wide conductive material that is sewn in and has a 1 MΩ resistor.
Spécifications
- Désignation:6' Coil Ground Cord Only,¼" Molded Female Snap