Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
1 µm, Plain back, 12"
  100497-238
Supplier #:  50354-10
Diamond Lapping Film, Electron Microscopy Sciences
  100497-238
 :  
Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
1 µm, Plain back, 12"
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis. Comes in 8" and 12" discs with plain back (P/B) or pressure sensitive adhesive back (PSA).
  • Granulométrie:
    1 µm
  • Taille:
    12"
  • Backing:
    Plain back