Diamond Lapping Film, Electron Microscopy Sciences | Avantor
Diamond Lapping Film, Electron Microscopy Sciences
15 µm, Plain back, 8"
Catalog # 100497-210
Fournisseur #:  50350-30
Diamond Lapping Film, Electron Microscopy Sciences
Catalog # 100497-210
Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
15 µm, Plain back, 8"
Price
96,05 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis. Comes in 8" and 12" discs with plain back (P/B) or pressure sensitive adhesive back (PSA).
Spécifications
  • Granulométrie:
    15 µm
  • Taille:
    8"
  • Backing:
    Plain back