Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks (Stainless Steel), 2"
  100496-978
 :  50152-05
Flat Mounting Blocks , Electron Microscopy Sciences
  100496-978
Flat Mounting Blocks , Electron Microscopy Sciences
Flat Mounting Blocks (Stainless Steel), 2"
192,31 $
Each
Stainless steel flat mounting blocks with the diameter of 25 mm (1") are used for lapping and polishing fixtures.
It is used for the models 150, 151 and 155.
  • Ø:
    2"
  • Désignation:
    Flat Mounting Blocks (Stainless Steel)