JEOL Strip Apertures, Electron Microscopy Sciences
Hitachi, Hole Sizes: 30, 20, 20, 70 µm; 41.0×4.0×0.10 mm
Catalog # 100496-796
 :  49211-02
JEOL Strip Apertures, Electron Microscopy Sciences
Catalog # 100496-796
JEOL Strip Apertures, Electron Microscopy Sciences
JEOL Strip Apertures, Electron Microscopy Sciences
Hitachi, Hole Sizes: 30, 20, 20, 70 µm; 41.0×4.0×0.10 mm
Price
$2,140.03
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Multihole strip aperture made from Platinum-Iridium (95:5).
Specifications
  • Désignation:
    Hitachi, Hole Sizes: 30, 20, 20, 70 µm; 41.0×4.0×0.10 mm