Micro Scissors, Castro-Viejo, Electron Microscopy Sciences
Castro-Viejo Micro Scissors, 51-12-30P, 130 mm (5"), Angled
  100494-422
 :  77892-A
Micro Scissors, Castro-Viejo, Electron Microscopy Sciences
  100494-422
Micro Scissors, Castro-Viejo, Electron Microscopy Sciences
Micro Scissors, Castro-Viejo, Electron Microscopy Sciences
Castro-Viejo Micro Scissors, 51-12-30P, 130 mm (5"), Angled
$542.83
Each
Castro Viejo Style PFC micro scissors with diamond dust.
  • Désignation:
    Castro-Viejo Micro Scissors, 51-12-30P
  • Longueur:
    130 mm (5")
  • Curvature:
    Angled