Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Wax Pen Replacement Tips
  100493-832
 :  72679-RT
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
  100493-832
 :  
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Wax Pen Replacement Tips
This pen helps to flatten and separate tissue sections and reduces compression in thin sections.
  • Powered by AA batteries
Delivery information: Available in two models: Wax Pen 1 is powered by one AA battery; Wax Pen 2 is powered by two AA batteries.
  • Désignation:
    Wax Pen Replacement Tips