Hot Pen - Wax Pen; A Tool for Separating Sections, Electron Microscopy Sciences
Product Details & Documents
This pen helps to flatten and separate tissue sections and reduces compression in thin sections.
- Powered by AA batteries
Delivery information: Available in two models: Wax Pen 1 is powered by one AA battery; Wax Pen 2 is powered by two AA batteries.
Spécifications
- Désignation:Wax Pen Replacement Tips