Count Up/Count Down Timer, Electron Microscopy Sciences
Product Details & Documents
Count up or down for 100 hours in intervals of 1 second.
- Alarm clock mode
- Its memory allows for the repeat of a cycle just completed
In the count down mode its unique feature tells how much time has elapsed after the alarm sounds.
Dimensions: 2³/₄×2³/₄×¹/₂"
Weight: 1.8 oz.
Certifications: With N.I.S.T. traceable certificate of compliance.
Delivery information: Comes complete with a built in stand, pocket clip, button cell battery, and magnet.
Specifications
- Désignation:Count up/count down timer certified